Orders LinkContactUsLinkComponent
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
공급사:   Avantor
J.T.Baker®, Cycle-Tap Sampler
공급사:  Avantor
Certificates
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
6 Options Available Below
Product Details & Documents
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
Product Family Options
제품 정보
  • Colour
    재고 여부
    Price
  • 9815-01
    Cycle-tap sampler, THF, high purity, low and ultra low water
    Cycle-tap sampler, THF, high purity, low and ultra low water
    Blue
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 9815-02
    Cycle-tap sampler, acetonitrile, high purity, low water
    Cycle-tap sampler, acetonitrile, high purity, low water
    Black
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 9815-03
    Cycle-tap sampler, methanol, high purity, low water
    Cycle-tap sampler, methanol, high purity, low water
    Orange
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 9815-04
    Cycle-tap sampler, DCM, high purity, low water
    Cycle-tap sampler, DCM, high purity, low water
    Green
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 9815-05
    Cycle-tap sampler
    Cycle-tap sampler
    Brown
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 9815-06
    Cycle-tap sampler, hexanes, high purity
    Cycle-tap sampler, hexanes, high purity
    Yellow