Orders LinkContactUsLinkComponent
J.T.Baker®, Liquid Level Indicators
J.T.Baker®, Liquid Level Indicators
CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
제품 번호 9803-03
공급사:   Avantor
J.T.Baker®, Liquid Level Indicators
제품 번호 9803-03
공급사:  Avantor
J.T.Baker®, Liquid Level Indicators
CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
Jump to:
Price
₩4,622,156
Log in 재고 확인
1 * 1 items
Product Details & Documents
These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
  • Signal can also be used to drive switching circuits
사양
  • 설명:
    CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system