Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
₩0
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Korea, Republic of
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
NDF volumetric solution
NDF volumetric solution, for analysis of animal feed (NF V18-122)
NDF volumetric solution, for analysis of animal feed (NF V18-122)
NDF volumetric solution, for analysis of animal feed (NF V18-122)
Catalog # 305320.2500
Supplier:
VWR Chemicals
CAS Number:
Certificates
Expand image
NDF volumetric solution, for analysis of animal feed (NF V18-122)
Catalog # 305320.2500
Supplier:
VWR Chemicals
CAS Number:
Certificates
Specifications
Pk:
2,5 L
Seq:
5096
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation