Orders LinkContactUsLinkComponent
VWR® Basic, 작업복
VWR® Basic, 작업복
VWR® Basic, 작업복
Supplier:  VWR Collection
Certificates
VWR® Basic, 작업복
8 Options Available Below
Product Details & Documents
칼라가 있는 이 일회용 작업복은 위험하지 않고 더러운 작업 환경에서 보호 기능을 제공하는 경량 원단인 스펀본드 폴리프로필렌(SPP) 부직포로 만들어졌습니다.
  • 칼라
  • 지퍼 잠금(한 방향)
  • 신축성 있는 손목
  • 천연 고무 라텍스 무첨가
무게: 40 g/m²
Packaging: 백당 1개, 한 케이스에 50개의 백으로 포장됩니다.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-9002
  • 113-9003
  • 113-9004
  • 113-9005
  • 113-9006
  • 113-9007
  • 113-9008
  • 113-9009
  • Garment application:
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • Weight:
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    의류 사이즈
    Availability
    Price
  • White
    L
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9002
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9003
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9004
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    3XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9005
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    L
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9006
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9007
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    XXL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9008
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    3XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9009
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²