Orders LinkContactUsLinkComponent
VWR® Basic, Frocks
VWR® Basic, Frocks
VWR® Basic, Frocks
VWR® Basic, Frocks
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
  • Press stud fastening
  • Elastic wrists
  • Raglan sleeves
  • Free from natural rubber latex
Weight: 40 g/m²
 : 1 piece per bag, 50 bags per case.
  • Catalog No:
  • 113-8995
  • 113-8996
  • 113-8998
  • 113-8999
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • Weight:
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
Product Family Options
  • Colour
    의류 사이즈
    길이
  • White
    L
    104 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8995
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XL
    108 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8996
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    L
    104 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8998
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    XL
    108 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8999
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²