Orders LinkContactUsLinkComponent
VWR® Basic, 후드 작업복
VWR® Basic, 후드 작업복
VWR® Basic, 후드 작업복
Supplier:  VWR Collection
Certificates
VWR® Basic, 후드 작업복
12 Options Available Below
Product Details & Documents
경량 원단인 부직포 스펀본드 폴리프로필렌(SPP)로 만들어진 일회용 후드 작업복은 위험하지 않고 더러운 작업 환경에서 보호 기능을 제공합니다.
  • 후드
  • 지퍼 잠금(한 방향)
  • 신축성있는 손목
  • 천연 고무 라텍스 무첨가
무게: 40 g/m²
Packaging: 백당 1개, 한 케이스에 50개의 백으로 포장됩니다.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-9014
  • 113-9015
  • 113-9016
  • 113-9017
  • 113-9018
  • 113-9019
  • 113-9020
  • 113-9021
  • 113-9022
  • 113-9023
  • 113-9024
  • 113-9025
  • Garment application:
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • Weight:
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    의류 사이즈
    Availability
    Price
  • White
    S
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9014
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    M
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9015
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    L
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9016
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9017
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9018
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    3XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9019
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    S
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9020
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    M
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9021
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    L
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9022
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9023
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    XXL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9024
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • Blue
    3XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9025
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²