Avantor® ACE® ChromSword, HPLC/UHPLC Columns, Method Development Kits
About this item
Solve U/HPLC method development challenges systematically and efficiently using Avantor® ACE® columns and ChromSword 5.1 Lite method development software. The Avantor® ACE® ChromSword Method Development Kit includes six alternative selectivity Avantor® ACE® phases, a copy of ChromSword 5.1 Lite offline with a six month activation licence and a lot of technical information and support.
- Ideal tool to investigate method development
- Excellent efficiency, reproducibility and column lifetime
- Columns with different mechanisms of interaction grouped in a kit to maximise selectivity and improve the likelihood of separating difficult or closely related analytes in mixtures
The ChromSword 5.1 Lite offline software in these method development kits is an introductory version of ChromSword, offering an extremely cost effective opportunity to explore the many benefits of the ChromSword 5.1 Lite software.
ChromSword 5.1 Lite is designed to help you develop and optimise U/HPLC separations more effectively and efficiently. On the basis of only a few experiments you can use the power of ChromSword to simulate numerous possible chromatographic solutions, looking at various columns and method conditions. This data can then be reviewed to identify and then optimise the separation most suitable to your needs. Typical method variables include organic modifier concentration, gradient profile, temperature, pH and much more. This empirical method development approach will provide you with the best possible separation for your analytes.
Furthermore, the six Avantor® ACE® columns in the kit have all been pre-calibrated by ChromSword and the data is stored within the library provided. This, together with data on physical chemistry properties, which is also provided, enables some predictive modelling (using the physical chemical properties of the molecular structures of your analytes) to be carried out. Elucidation of a purely theoretical chromatogram is, therefore, possible with this approach, and although it will be less precise than the 'hands-on', empirical approach, it may provide a quick indication of the direction in which to focus your efforts. However, whatever a theoretical approach may indicate, it is always recommended that various column selectivities are screened. With these kits, you have a choice of up to six different Avantor® ACE® column selectivities to explore.
The ChromSword licence included with the kit will provide you with a full six months of method development investigation time. After this you can choose to extend or upgrade the software licence.
Delivery information: Avantor® ACE® ChromSword Method Development Kits contain Avantor® ACE® C18, Avantor® ACE® C18-AR, Avantor® ACE® C18-PFP, Avantor® ACE® CN-ES, Avantor® ACE® C18-Amide and Avantor® ACE® SuperC18 columns, ChromSword 5.1 Lite Offline version. Kits with various dimensions and particle sizes are available. All columns in the kit are the same dimensions and particle size.
35 Options Available Below
Product Details & Documents
Solve U/HPLC method development challenges systematically and efficiently using Avantor® ACE® columns and ChromSword 5.1 Lite method development software. The Avantor® ACE® ChromSword Method Development Kit includes six alternative selectivity Avantor® ACE® phases, a copy of ChromSword 5.1 Lite offline with a six month activation licence and a lot of technical information and support.
- Ideal tool to investigate method development
- Excellent efficiency, reproducibility and column lifetime
- Columns with different mechanisms of interaction grouped in a kit to maximise selectivity and improve the likelihood of separating difficult or closely related analytes in mixtures
The ChromSword 5.1 Lite offline software in these method development kits is an introductory version of ChromSword, offering an extremely cost effective opportunity to explore the many benefits of the ChromSword 5.1 Lite software.
ChromSword 5.1 Lite is designed to help you develop and optimise U/HPLC separations more effectively and efficiently. On the basis of only a few experiments you can use the power of ChromSword to simulate numerous possible chromatographic solutions, looking at various columns and method conditions. This data can then be reviewed to identify and then optimise the separation most suitable to your needs. Typical method variables include organic modifier concentration, gradient profile, temperature, pH and much more. This empirical method development approach will provide you with the best possible separation for your analytes.
Furthermore, the six Avantor® ACE® columns in the kit have all been pre-calibrated by ChromSword and the data is stored within the library provided. This, together with data on physical chemistry properties, which is also provided, enables some predictive modelling (using the physical chemical properties of the molecular structures of your analytes) to be carried out. Elucidation of a purely theoretical chromatogram is, therefore, possible with this approach, and although it will be less precise than the 'hands-on', empirical approach, it may provide a quick indication of the direction in which to focus your efforts. However, whatever a theoretical approach may indicate, it is always recommended that various column selectivities are screened. With these kits, you have a choice of up to six different Avantor® ACE® column selectivities to explore.
The ChromSword licence included with the kit will provide you with a full six months of method development investigation time. After this you can choose to extend or upgrade the software licence.
Delivery information: Avantor® ACE® ChromSword Method Development Kits contain Avantor® ACE® C18, Avantor® ACE® C18-AR, Avantor® ACE® C18-PFP, Avantor® ACE® CN-ES, Avantor® ACE® C18-Amide and Avantor® ACE® SuperC18 columns, ChromSword 5.1 Lite Offline version. Kits with various dimensions and particle sizes are available. All columns in the kit are the same dimensions and particle size.
Specifications for Product Family
Specifications
- Catalog No:
- MDKCS-17-0502U
- MDKCS-17-1002U
- MDKCS-17-0503U
- MDKCS-17-1003U
- MDKCS-2-0502U
- MDKCS-2-1002U
- MDKCS-2-1502U
- MDKCS-2-0503U
- MDKCS-2-1003U
- MDKCS-2-1503U
- MDKCS-2-0546U
- MDKCS-2-1046U
- MDKCS-2-1546U
- MDKCS-3-0502U
- MDKCS-3-1002U
- MDKCS-3-1502U
- MDKCS-3-2502U
- MDKCS-3-0503U
- MDKCS-3-1003U
- MDKCS-3-1503U
- MDKCS-3-0546U
- MDKCS-3-1046U
- MDKCS-3-1546U
- MDKCS-5-0502U
- MDKCS-5-1002U
- MDKCS-5-1502U
- MDKCS-5-2502U
- MDKCS-5-0503U
- MDKCS-5-1003U
- MDKCS-5-1503U
- MDKCS-5-2503U
- MDKCS-5-0546U
- MDKCS-5-1046U
- MDKCS-5-1546U
- MDKCS-5-2546U
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Product Family Options
Avantor® ACE® ChromSword method development kits, 1,7 µm
Product Information
- 컬럼 형식Phase기공 크기입자 크기내부 직경길이Packing materialAvailabilityPrice
- Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å1,7 µm2,10 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-17-0502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å1,7 µm2,10 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-17-1002UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å1,7 µm3,00 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-17-0503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å1,7 µm3,00 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-17-1003UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Avantor® ACE® ChromSword method development kits, 2 µm
Product Information
- 컬럼 형식Phase기공 크기입자 크기내부 직경길이Packing materialAvailabilityPrice
- Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm2,10 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-0502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm2,10 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-1002UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm2,10 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-1502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm3,00 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-0503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm3,00 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-1003UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm3,00 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-1503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm4,60 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-0546UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm4,60 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-1046UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å2 µm4,60 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-2-1546UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Avantor® ACE® ChromSword method development kits, 3 µm
Product Information
- 컬럼 형식Phase기공 크기입자 크기내부 직경길이Packing materialAvailabilityPrice
- Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm2,10 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-0502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm2,10 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-1002UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm2,10 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-1502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm2,10 mm250 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-2502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm3,00 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-0503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm3,00 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-1003UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm3,00 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-1503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm4,60 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-0546UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm4,60 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-1046UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å3 µm4,60 mm150 mmSilicaLog in for availability1 * 1 KITSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-3-1546UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Avantor® ACE® ChromSword method development kits, 5 µm
Product Information
- 컬럼 형식Phase기공 크기입자 크기내부 직경길이Packing materialAvailabilityPrice
- Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm2,10 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-0502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm2,10 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-1002UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm2,10 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-1502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm2,10 mm250 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-2502UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm3,00 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-0503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm3,00 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-1003UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm3,00 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-1503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm3,00 mm250 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-2503UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm4,60 mm50 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-0546UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm4,60 mm100 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-1046UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm4,60 mm150 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-1546UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Method development kitC18, C18-AR, C18-PFP, CN-ES, C18-Amide, SuperC18100 Å5 µm4,60 mm250 mmSilicaLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MDKCS-5-2546UEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact