Orders LinkContactUsLinkComponent
3중 디스플레이 온도계, Traceable
3중 디스플레이 온도계, Traceable
Supplier:   VWR Collection
3중 디스플레이 온도계, Traceable
Supplier:  VWR Collection
Certificates
3중 디스플레이 온도계, Traceable
3중 디스플레이 온도계, Traceable
2 Options Available Below
Product Details & Documents
These digital thermometers are suitable for monitoring of temperatures in freezers, water baths, heating blocks, incubators and refrigerators. The bottle-enclosed, temperature-buffered sensor is insulated from transient temperature changes. Micro cable allows refrigerator doors to close on it.
  • Maximum/minimum monitors high and low readings overnight, at the weekend or for any time period
  • Programmable alarm sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • Visual and audible signals continue even if temperature returns to non alarm range
Waterproof probe (Lר: 19×8 mm) or bottle-buffered sensor, with 3 metre cable.
Conforms to ISO 17025 and is traceable to NIST.
Patented unit fullfils all CDC thermometer and vaccine thermometer requirements.
Certifications: Compliance with ISO/IEC 17025, ANSI/NCSL Z541-1 and 9001. Traceability to NIST. Patented unit fulfils all CDC thermometer and vaccine thermometer requirements.
Delivery information: Supplied with Traceable® certificate of calibration, flip stand, wall bracket, VELCRO mounting strips and batteries.
Product Family Options
Product Information
  • Range
    Accuracy
    해상도
    Probe
    W×D×H
    Weight
    Availability
    Price
  • –50…+70 °C
    ±1,0
    1,0 °C
    Waterproof
    70×20×120 mm
    113 g
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • −50…+70 °C
    ±1,0
    1,0 °C
    Bottle-buffered sensor
    70×20×120 mm
    113 g