Avantor® Genesis, HPLC Columns, AQ
: Avantor
Avantor® Genesis phases are based on high purity metal-free spherical silica. They are suitable for the analysis of a wide range of analytes.
- Good peak shape and reproducibility
- Long column lifetime
- pH stability 1 to 10
- Catalog No:
- FM15951E
- FH1951-2/N
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 15%
- Endcapped:
- Yes
- pH range:
- 1 - 10
- Surface area:
- 300 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Frequently Bought Together