Vydac 208MS C8, HPLC columns
공급사: Avantor
인증서
이 제품에 관하여
Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules. A proprietary surface treatment and bonding process give Vydac MS columns unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
- Five reversed phase chemistries
- Excellent peak shape with little or no TFA
- High protein recoveries make scale-up easy
Lower hydrophobicity which is better for larger biomolecules. Ideal for biomolecules with 5 to 10 k MW.
Vydac is a registered trademark of W.R.Grace and Co.
사양
- Catalog No:
- 208MS52
- 208MS54
- 208MS5415
- 208MS5215
- 208MS5210
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 5%
- 5%
- 5%
- 5%
- 5%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L7
- L7
- L7
- L7
- L7
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)