VWR®EZwaste™, Solvent waste system
These solvent waste system is designed to be an affordable solution for HPLC liquid waste disposal.
- Activated carbon filter traps volatile organic compounds
- Prevents chemicals from being inhaled
- Closed loop design
Manufactured to safely collect waste from two HPLC machines. Features an activated carbon filter which traps volatile organic compounds from being released into the air. Prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapour wastes. The solvent waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.
: Each system includes a carboy, two 83B versatile caps (one open top for waste collection and one closed top for safe transport), six ports for 3,2 mm Ext.Ø waste tubes, one port for a carbon exhaust filter, one carbon exhaust filter and plugs for unused ports.
These solvent waste system is designed to be an affordable solution for HPLC liquid waste disposal.
- Activated carbon filter traps volatile organic compounds
- Prevents chemicals from being inhaled
- Closed loop design
Manufactured to safely collect waste from two HPLC machines. Features an activated carbon filter which traps volatile organic compounds from being released into the air. Prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapour wastes. The solvent waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.
: Each system includes a carboy, two 83B versatile caps (one open top for waste collection and one closed top for safe transport), six ports for 3,2 mm Ext.Ø waste tubes, one port for a carbon exhaust filter, one carbon exhaust filter and plugs for unused ports.
Product Family Options
- 설명용량
- Solvent waste carboy kit, bundle, 80 mm cap size4 L1 * 1 items₩1,980,497
- Solvent waste carboy kit, bundle, 80 mm cap size5 L1 * 1 items₩2,084,051
- Solvent waste carboy kit, bundle, 80 mm cap size10 L1 * 1 items₩2,399,128
- Solvent waste carboy kit, bundle, 80 mm cap size20 L1 * 1 items₩2,419,167