VWR®, Filter forceps
114 mm
제품 번호 VWRU89259-954
공급사: VWR Collection
Jump to:
Product Details & Documents
These stainless steel forceps with black PP non-slip finger grips are ideal for handling membranes.
사양
- 길이:114 mm
These stainless steel forceps with black PP non-slip finger grips are ideal for handling membranes.