Orders LinkContactUsLinkComponent
VWR®, Cryoboxes, PP, Flat-Packed
VWR®, Cryoboxes, PP, Flat-Packed
Supplier:   VWR Collection
VWR®, Cryoboxes, PP, Flat-Packed
Supplier:  VWR Collection
Certificates
VWR®, Cryoboxes, PP, Flat-Packed
VWR®, Cryoboxes, PP, Flat-Packed
6 Options Available Below
Product Details & Documents
Made of corrugated or thin film PP.
  • Easy self-assembly boxes shipped flat-packed
  • More durable than cardboard boxes as they are not susceptible to moisture
  • Significantly reduces the chance of mould
  • Dividers included with the boxes
  • Store down to temperatures of –80 °C
Designed with sustainability in mind, they are made from either specially formulated corrugated or thin film polypropylene. With superior durability, polypropylene outlasts the cardboard box alternative that is susceptible to softening and producing dust, and can be frozen and thawed multiple times without a problem. These cryoboxes are supplied 'flat-packed', which eliminates the hassle of stacking, and reduces the amount of space needed for storage.
Product Family Options
Boxes, thin film PP
Product Information
  • Compartments
    Colour
    For tubes
    Well Ø
    W×D×H ext.
    Availability
    Price
  • 81
    White
    1,5/2,0 ml
    12,1
    125×125×49 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
Boxes, corrugated PP
Product Information
  • Compartments
    Colour
    For tubes
    Well Ø
    W×D×H ext.
    Availability
    Price
  • 144
    Natural
    0,2 ml
    7,9
    133×130×30 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 81
    Natural
    0,5 ml
    12,0
    133×130×42 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 25
    Natural
    5 ml
    22,6
    133×130×75 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 36
    Natural
    15 mm
    19,8
    145×147×120 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 16
    Natural
    50 ml
    31,6
    145×147×120 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items