Vydac Denali C18, HPLC columns
공급사: Avantor
인증서
이 제품에 관하여
Vydac Denali (238DE) is a 120 Å C18 bonded phase with high carbon coverage, suitable for the analysis of both acidic and basic analytes. It has applications for small molecule analyses of interest to pharmaceutical and environmental laboratories.
- High retentivity
- LC-MS of small molecules
- Fully scalable from capillary to process
* Use with holder GCH10 and column coupler HI-081.
** Use with holder 80101/N and column coupler HI-081.
Vydac is a registered trademark of W.R.Grace and Co.
사양
- Catalog No:
- 238DE5415
- 238DE54
- 238DE5410
- 238DE1022
- 238GD54DE/N
- 238GD52DE/N
- 238DE52215
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 20%
- 20%
- 20%
- 20%
- 20%
- 20%
- 20%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)