J.T.Baker® BAKERBOND Speedisk® C18 PolarPlus®, SPE Columns
Catalog # 8153-04
Supplier: Avantor
Specifications
- 용량:3
- Seq:02
- Bed weight:50 mg
- Phase:Octadecyl (C18) PolarPlus
- Pk:1
Specifications
About this item
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
- Run times up to nine times faster than conventional SPE columns
- Unique laminar configuration
- Silica based
General applications: Non ionic, basic, non polar to polar analytes.
These SPE columns can be used in reversed phase mode.