Avantor® Alltima, HPLC Columns, Amino
공급사: Avantor
인증서
이 제품에 관하여
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L8
General purpose amino suitable for normal or reversed phase use. Recommended for carbohydrate analysis or as a weak anion exchanger.
사양
- Catalog No:
- HICH88217
- 88217RP
- HICH88218
- AL-5NH-250DF
- HICH81183
- HICH81190
- 96085/N
- HICH88205
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- Surface area:
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- USP code:
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)