Orders LinkContactUsLinkComponent
VWR® Traceable®, Large digit 3중 디스플레이 온도계
VWR® Traceable®, Large digit 3중 디스플레이 온도계
Supplier:   VWR Collection
VWR® Traceable®, Large digit 3중 디스플레이 온도계
Supplier:  VWR Collection
Certificates
VWR® Traceable®, Large digit 3중 디스플레이 온도계
VWR® Traceable®, Large digit 3중 디스플레이 온도계
2 Options Available Below
Product Details & Documents
These jumbo size display thermometers are suitable for monitoring freezers, water baths, heating blocks, incubators and refrigerators. A bottle insulates the sensor from transient temperature changes when the refrigerator door is opened. Solution is GRAS (Generally Recognised As Safe) by the FDA (Food and Drug Administration).
  • Triple display: Maximum/minimum and current temperatures
  • Out of range alarm programmable in 1,0 °C increments
Bottle probe in non toxic glycol solution or waterproof bullet probe (20 mm).
Bottle probe sensor: Temperature-buffered glycol bottle probe mimics solution. Bottle and solution insulate sensor from rapid changes, as an example, when a refrigerator door is opened. Solution mimics temperature of stored samples. Probe is sealed in a miniature bottle filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration).
Bullet probe sensor: Small sized plastic probe designed for use in air and liquids, the sensor and cable may be completely immersed.
Certifications: Traceability to NIST. Fulfils CDC and vaccine thermometer requirements. Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with Traceable® certificate of calibration, slot for wall mounting, VELCRO® and magnetic strips to attach to any surface, AAA battery and 3 metre cable.
Product Family Options
Product Information
  • 설명
    Range
    Accuracy
    해상도
    Probe
    W×D×H
    Weight
    Availability
    Price
  • Thermometer with bottle probe sensor
    −50…+70 °C
    ±1,0
    0,1 °C
    Bottle
    99×111×22 mm
    113 g
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Thermometer with waterproof bullet probe sensor
    −50…+70 °C
    ±1,0
    0,1 °C
    Waterproof bullet
    99×111×22 mm
    113 g