VWR® PESTINORM®, iso-Butyl Methyl Ketone, Secondary Reference Standard for GC
Catalog # 85673.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- 포장 유형:Glass bottle
- Pk:100 mL
- 설명:iso-Butyl methyl ketone standard for GC
Specifications
About this item
A secondary GC solvent reference standard for various chromatographic and analytical applications.
- Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
- Traceability data to specific current lot of primary standard
- Completely synthetic and typically over 99,9% pure by GC-FID