VWR® PESTINORM®, Dimethylsulphoxide (DMSO), Secondary Reference Standard for GC
85663.180
:
- 포장 유형:Glass bottle
- Pk:100 mL
- 설명:Dimethylsulphoxide (DMSO) standard for GC
A secondary GC solvent reference standard for various chromatographic and analytical applications.
- Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
- Traceability data to specific current lot of primary standard
- Completely synthetic and typically over 99,9% pure by GC-FID