Orders LinkContactUsLinkComponent
LiChrosorb® RP-18, HPLC Columns
LiChrosorb® RP-18, HPLC Columns
   Sustainable Options Available
Supplier:   Avantor
LiChrosorb® RP-18, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
LiChrosorb® RP-18, HPLC Columns
15 Options Available Below
Product Details & Documents
These columns with high purity base deactivated porous silica are ideal for the separation of basic molecules.
  • Irregular porous silica
  • High efficiency and complimentary phase selectivity
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • LIRP18-10-10C5
  • LIRP181010C5SK
  • LIRP18-5-10C5
  • LIRP1810125AF
  • LIRP18-5-125AF
  • LIRP18-5-150AF
  • LIRP18-10250AF
  • LIRP18-5-250AF
  • LIRP1810300AF
  • LIRP18-10-100A
  • LIRP18-5-125A
  • LIRP18-5-150A
  • LIRP18-10-200A
  • LIRP18-10-250A
  • LIRP18-5-250A
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • 21,0%|16,2%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • 350 m²/g|300 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Family Options
LiChrosorb® Guard cartridges
Product Information
  • Phase
    컬럼 형식
    기공 크기
    입자 크기
    내부 직경
    길이
    Availability
    Price
  • RP-18
    Guard cartridge
    100 Å
    10 µm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-10-10C5
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    Guard cartridge
    100 Å
    10 µm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP181010C5SK
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    Guard cartridge
    100 Å
    5 µm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-5-10C5
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
LiChrosorb® Analytical columns
Product Information
  • Phase
    컬럼 형식
    기공 크기
    입자 크기
    내부 직경
    길이
    Availability
    Price
  • RP-18
    100 Å
    10 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP1810125AF
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    5 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-5-125AF
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    5 µm
    4,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-5-150AF
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    10 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-10250AF
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    5 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-5-250AF
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    10 µm
    4,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP1810300AF
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    10 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-10-100A
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    5 µm
    4,60 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-5-125A
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    5 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-5-150A
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    10 µm
    4,60 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-10-200A
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    10 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-10-250A
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • RP-18
    100 Å
    5 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    LIRP18-5-250A
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    21,0%|16,2%
    Endcapped
    No
    Surface area
    350 m²/g|300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)