Orders LinkContactUsLinkComponent
VWR®, Bottle assemblies, PC, 멸균, 일회용
VWR®, Bottle assemblies, PC, 멸균, 일회용
VWR®, Bottle assemblies, PC, 멸균, 일회용
공급사:  VWR Collection
Certificates
VWR®, Bottle assemblies, PC, 멸균, 일회용
VWR®, Bottle assemblies, PC, 멸균, 일회용
6 Options Available Below
Product Details & Documents
Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They comes in a comprehensive size selection with tubing that is completely weldable/sealable.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-6
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Product Family Options
제품 정보
  • 용량
    캡 크기
    Tubing connection
    Tubing length
    Tubing material
    Tubing size
    재고 여부
    Price
  • 125
    38-430
    Port 1: PP Tube Plug
    Port 2: 25 mm Hydrophobic PE Vent Filter
    Port 1: 24"
    Port 2: 3"
    C-Flex
    Port 1: 1/8 x 1/4"
    Port 2: 1/8 x 1/4"
    Log in 재고 확인
    Log in 재고 확인
    1 * 6 items
    1 * 6 items
  • 250
    38-430
    Port 1: PP Tube Plug
    Port 2: 25 mm Hydrophobic PE Vent Filter
    Port 1: 24"
    Port 2: 3"
    C-Flex
    Port 1: 1/8 x 1/4"
    Port 2: 1/8 x 1/4"
    Log in 재고 확인
    Log in 재고 확인
    1 * 6 items
    1 * 6 items
  • 500
    38-430
    Port 1: PP Tube Plug
    Port 2: 25 mm Hydrophobic PE Vent Filter
    Port 1: 24"
    Port 2: 3"
    C-Flex
    Port 1: 1/8 x 1/4"
    Port 2: 1/8 x 1/4"
    Log in 재고 확인
    Log in 재고 확인
    1 * 6 items
    1 * 6 items
  • 1000
    38-430
    Port 1: PP Tube Plug
    Port 2: 25 mm Hydrophobic PE Vent Filter
    Port 1: 24"
    Port 2: 3"
    C-Flex
    Port 1: 1/8 x 1/4"
    Port 2: 1/8 x 1/4"
    Log in 재고 확인
    Log in 재고 확인
    1 * 3 items
    1 * 3 items
  • 2000
    38-430
    Port 1: PP Tube Plug
    Port 2: 50 mm PTFE Vent Filter
    Port 1: 24"
    Port 2: 3"
    C-Flex
    Port 1: 1/8 x 1/4"
    Port 2: 1/4 x 3/8"
    Log in 재고 확인
    Log in 재고 확인
    1 * 2 items
    1 * 2 items
  • 2000
    53B
    Port 1: PP Tube Plug
    Port 2: 50 mm PTFE Vent Filter
    Port 3: PP Tube Plug
    Port 1: 12"
    Port 2: 3"
    Port 3: 12"
    C-Flex
    Port 1: 3/8 x 1/2"
    Port 2: 1/4 x 3/8"
    Port 3: 3/8 x 1/2"
    Log in 재고 확인
    Log in 재고 확인
    1 * 2 items
    1 * 2 items