Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution USP Test Solution (TS)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Merck Index:12,08844
Boiling Pt:100 °C (1013 hPa)
Melting Pt:48 °C
Storage Temperature:Ambient
EINECS:231-867-5
UN:0000
Frequently Bought Together