Orders LinkContactUsLinkComponent
VWR®, Micro Spatulas
VWR®, Micro Spatulas
공급사:   VWR Collection
VWR®, Micro Spatulas
공급사:  VWR Collection
Certificates
VWR®, Micro Spatulas
VWR®, Micro Spatulas
2 Options Available Below
Product Details & Documents
Stainless steel. These micro spatulas are ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
  • Rustproof material
  • With non stick surfaces
  • Mirror finish
They feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.
Product Family Options
제품 정보
  • 길이
    재고 여부
    Price
  • 165 mm
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items - ₩20,142
    ₩20,142
    1 * 1 items
       ₩20,142
  • 203 mm
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items - ₩20,142
    ₩20,142
    1 * 1 items
       ₩20,142