VWR®, Ring Weights for Flasks, C-Shaped
Flasks 500 - 2000 ml, 56, 680 g
제품 번호 VWRU47735-598
공급사: VWR Collection
Jump to:
Product Details & Documents
Lead core, with Vikem® vinyl coating. These lead rings fit neatly around the neck of filtering flasks with side-arm tubing connections without disconnecting the tubing.
- Stabilise vessels in water baths or on the benchtop
- Open ring is flexible enough to be pulled apart to accommodate a wider vessel
사양
- Weight:680 g
- Seq:03
- For:Flasks 500 - 2000 ml
- 외부 직경:56