VWR®, Self Seal Sterilisation Pouches with Indicator
공급사: VWR Collection
인증서
이 제품에 관하여
Sterilisation pouches are made of surgical grade paper and blue polypropylene/polyester film. Color-coded indicators on every pouch.
- Easy-to-open lip closure
- Prefold self-adhesive strip
- Flat seals ensure seal integrity
Frequently Bought Together