Orders LinkContactUsLinkComponent
VWR®, Spatulas
Supplier:   VWR Collection
VWR®, Spatulas
Supplier:  VWR Collection
Certificates
VWR®, Spatulas
3 Options Available Below
Product Details & Documents
Stainless steel spatulas with wooden handle. Economical tools for handling, transferring, blending, or collecting chemical and biological samples.
  • Sturdy and durable
  • Stainless steel construction
  • Multiple sizes allow for varied laboratory applications
Product Family Options
Product Information
  • 설명
    길이
    Blade length
    Availability
    Price
  • Standard blade
    127
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩33,892
    ₩33,892
    1 * 1 items
       ₩33,892
  • Standard blade
    304 mm
    152
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩42,695
    ₩42,695
    1 * 1 items
       ₩42,695
  • Standard blade
    305
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩85,538
    ₩85,538
    1 * 1 items
       ₩85,538