VWR®, Low Temperature Slide Box, Polycarbonate
제품 번호 631-1143
공급사: VWR Collection
사양
- No. of slides:25
- Colour:Blue
- 설명:Slide box
- Material:PC
- Pk:1
사양
이 제품에 관하여
This slide box can accommodate up to 25 microscope slides.
- Numbered index sheet inside the box
- Cover lid to secure the slides
- Easy to use and store