Orders LinkContactUsLinkComponent
VWR®, Cryobox, Light-Sensitive
10×10, Microtubes up to 2,0 ml, Black, 140×140×60 mm
제품 번호 VWRU80080-597
공급사:   VWR Collection
VWR®, Cryobox, Light-Sensitive
제품 번호 VWRU80080-597
공급사:  VWR Collection
VWR®, Cryobox, Light-Sensitive
VWR®, Cryobox, Light-Sensitive
10×10, Microtubes up to 2,0 ml, Black, 140×140×60 mm
Jump to:
Price
가격 문의
Log in 재고 확인
1 * 1 items
Product Details & Documents
PP, black.
  • The reinforced seal, triple hinges and locking claps keeps samples secure
  • Alpha-numeric grid on the exterior and interior of the box
  • Stacking rings and a matte surface suitable for printing and labelling
Ideal for light-sensitive specimens and suitable for micro-tubes and other cryogenic vials.
사양
  • Compartments:
    10×10
  • For:
    Microtubes up to 2,0 ml
  • W×D×H ext.:
    140×140×60 mm
  • Colour:
    Black