Formaldehyde 36,5 - 38,0% (by acidimetry) in aqueous solution stabilised, AR® ACS for histology, Macron Fine Chemicals™
Catalog # 5016-26
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- 포장 유형:PE drum, black
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Store between 15° C and 30° C:36.5 - 38.0 %
- ACS - Assay (HCHO) (by acidimetry):36.5 - 38.0 %
- ACS - Color (APHA):≤10
- ACS - Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤5 ppm
- Identification A:Passes Test
- Identification B:Passes Test
- ACS - Iron (Fe):≤5 ppm
- ACS - Methanol (CH₃OH):10 - 12 %
- ACS - Residue after Ignition:≤0.005 %
- ACS - Sulfate (SO₄):≤0.002 %
- ACS - Titrable Acid (meq/g):≤0.006
Specification Test Results
About this item
:
:30.03 g/mol
Boiling Pt:-19.3 °C (1013 hPa)
EINECS:200-001-8
ADR:8,III
:Management of Change (MOC) category = N
:Stabilised with methanol 10-15 %
UN:2209
:GHS06
:GHS05
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274