Orders LinkContactUsLinkComponent
VWR®, Sample library tubes
VWR®, Sample library tubes
공급사:   VWR Collection
VWR®, Sample library tubes
공급사:  VWR Collection
Certificates
VWR®, Sample library tubes
VWR®, Sample library tubes
2 Options Available Below
Product Details & Documents
PP, clear.
  • Autoclavable and freezable (–80 °C)
  • Printed with an alpha numeric grid that helps to find a sample easily
These 1,2 ml sample library tubes are supplied in a 96 tube format for storage of specimens. The rack is designed to meet SBS standards and can be used in robotic applications.
납품 정보: Supplied in 8×12 racks with clear, snug fit lid.
Product Family Options
Tubes in racks
제품 정보
  • Colour
    재고 여부
    Price
  • VWRU82006-698
    Individual tubes
    Individual tubes
    Clear
    Log in 재고 확인
    Log in 재고 확인
    1 * 960 items
    1 * 960 items - ₩349,272
    ₩349,272
    1 * 960 items
       ₩349,272
  • VWRU93000-040
    Individual tubes, sterile
    Individual tubes, sterile
    Clear
    Log in 재고 확인
    Log in 재고 확인
    1 * 960 items
    1 * 960 items - ₩254,369
    ₩254,369
    1 * 960 items
       ₩254,369
Accessories
제품 정보
  • 제품 번호
    재고 여부
    Price
  • 12-well closure strips, sterile
    12-well closure strips, sterile
    VWRU93000-048
    Log in 재고 확인
    1 * 960 items
제품 정보
  • 제품 번호
    재고 여부
    Price
  • 8-well closure strips
    8-well closure strips
    VWRU82006-694
    Log in 재고 확인
    1 * 125 items - ₩100,256