VWR®, Draining Racks
Drying rack, 450×110×630 mm
제품 번호 VWRU60996-012
공급사: VWR Collection
Jump to:
Product Details & Documents
PS.
- Lightweight and versatile
- With 76 mm draining ledge
- Pegs snap out easily
This rack accommodates cylinders, flasks and test tubes.
사양
- 설명:Drying rack
- W×D×H:450×110×630 mm