Orders LinkContactUsLinkComponent
VWR®, Cork Stoppers
31 mm, 27 mm, 33 mm
  VWRU23420-366
VWR®, Cork Stoppers
  VWRU23420-366
VWR®, Cork Stoppers
VWR®, Cork Stoppers
31 mm, 27 mm, 33 mm
₩1,773,992
1 * 100 items
XXXX grade cork.
  • Tapered regular length
  • Selected grade of fine textured corks
  • Ø bottom:
    27 mm
  • Ø top:
    33 mm
  • 높이:
    31 mm
  • Seq:
    04