Avantor® ACE® UltraCore SuperC18, HPLC/UHPLC Columns, 5 µm
공급사: Avantor
인증서
이 제품에 관하여
Avantor® ACE® UltraCore SuperC18 solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. These stainless steel columns combine high efficiency separations with low column back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and effectively eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability.
- Ideal for demanding MS applications
- Excellent peak shape and phases stability with extended pH
- Highly inert, ultra-low bleed, designed for use with LC-MS compatible buffers
- Available with 2,5 and 5 µm particle sizes and microbore to analytical dimensions
Maximum recommended pH: 1,5 to 11,0
Max. pressure: 1000 bar
사양
- Catalog No:
- CORE-5A-1002U
- CORE-5A-2546U
- CORE-5A-0546U
- CORE-5A-2503U
- CORE-5A-0503U
- CORE-5A-2502U
- CORE-5A-1546U
- CORE-5A-0502U
- CORE-5A-1046U
- CORE-5A-1003U
- CORE-5A-0346U
- CORE-5A-1503U
- CORE-5A-1502U
- CORE-5A-1246U
- CORE-5A-0302U
- CORE-5A-7502U
- CORE-5A-0202U
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- 7,0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- pH range:
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- 1,5 - 11
- Surface area:
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)