Orders LinkContactUsLinkComponent
VWR®, 평량 보트, 종이
VWR®, 평량 보트, 종이
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 평량 보트, 종이
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 평량 보트, 종이
6 Options Available Below
Product Details & Documents
VWR® 종이 경량 보트는 첫 번째 친환경 실험실 소모품 제품입니다.
  • 폴리스티렌 평량 보트처럼 내용물을 제어하여 표준 평면 평량 보트보다 다루기 쉬움
  • FDA 21 CFR 176:170을 충족하고, 수분 및 지방이 많은 식품과 접촉하는 종이의 성분 및 건조 식품 접촉용 FDA 21 CFR 176.170을 충족하는 기름 방지 종이로 제작됨
  • 최대 210 ºC의 열 어플리케이션에 사용 가능
Eco-InEco-OUT 둘 다 만족합니다.
Eco-In : FSC® 인증을 받았으며 보트를 만드는 데 사용되는 종이는 목재 제품들로 만들어졌습니다.
Eco-out : 도시 폐기물 유량 에서 자연적으로 생분해됩니다.
Specifications for Product Family
Specifications
  • Catalog No:
  • 611-3258
  • 611-3256
  • 611-3259
  • 611-3257
  • 611-3625
  • 611-3624
  • Environmentally Preferable:
  • Waste Reducing|Sustainable Materials
  • Waste Reducing|Sustainable Materials
  • Waste Reducing|Sustainable Materials
  • Waste Reducing|Sustainable Materials
  • Sustainable Materials|Sustainable Packaging
  • Sustainable Materials
Specifications
Product Family Options
Product Information
  • 설명
    W×D×H
    Colour
    Availability
    Price
  • FSC® Certified
    35×35×15 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Waste Reducing|Sustainable Materials
    More
    Specifications
    Cat. No.
    611-3258
    Environmentally Preferable
    Waste Reducing|Sustainable Materials
      Waste Reducing|Sustainable Materials
  • FSC® Certified
    35×35×15 mm
    Brown
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Waste Reducing|Sustainable Materials
    More
    Specifications
    Cat. No.
    611-3256
    Environmentally Preferable
    Waste Reducing|Sustainable Materials
      Waste Reducing|Sustainable Materials
  • FSC® Certified
    45×45×15 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Waste Reducing|Sustainable Materials
    More
    Specifications
    Cat. No.
    611-3259
    Environmentally Preferable
    Waste Reducing|Sustainable Materials
      Waste Reducing|Sustainable Materials
  • FSC® Certified
    45×45×15 mm
    Brown
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Waste Reducing|Sustainable Materials
    More
    Specifications
    Cat. No.
    611-3257
    Environmentally Preferable
    Waste Reducing|Sustainable Materials
      Waste Reducing|Sustainable Materials
  • FSC® Certified
    92×92×25 mm
    White
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
    Specifications:
      Sustainable Materials|Sustainable Packaging
    More
    Specifications
    Cat. No.
    611-3625
    Environmentally Preferable
    Sustainable Materials|Sustainable Packaging
      Sustainable Materials|Sustainable Packaging
  • FSC® Certified
    92×92×25 mm
    Brown
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
    Specifications:
      Sustainable Materials
    More
    Specifications
    Cat. No.
    611-3624
    Environmentally Preferable
    Sustainable Materials
      Sustainable Materials