VWR®, 작업복, SPP
공급사: VWR Collection
이 제품에 관하여
Breathable overalls made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- With collar
- Two-way nylon zip
- Elasticated cuffs and ankles
Weight: 40 g/m².
Colour: White.
4 Options Available Below
Product Details & Documents
Breathable overalls made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- With collar
- Two-way nylon zip
- Elasticated cuffs and ankles
Weight: 40 g/m².
Colour: White.
Specifications for Product Family
사양
- Catalog No:
- 113-8311
- 113-8312
- 113-8313
- 113-8314
- Model:
- With collar
- With collar
- With collar
- With collar
- Garment application:
- General Purpose|Industrial
- General Purpose|Industrial
- General Purpose|Industrial
- General Purpose|Industrial
- Weight:
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
사양
Product Family Options
제품 정보
- Colour의류 사이즈재고 여부Price
- 사양:
More
Specifications
Cat. No.113-8311ModelWith collarGarment applicationGeneral Purpose|IndustrialWeight40 g/m² - 사양:
More
Specifications
Cat. No.113-8312ModelWith collarGarment applicationGeneral Purpose|IndustrialWeight40 g/m² - 사양:
More
Specifications
Cat. No.113-8313ModelWith collarGarment applicationGeneral Purpose|IndustrialWeight40 g/m² - 사양:
More
Specifications
Cat. No.113-8314ModelWith collarGarment applicationGeneral Purpose|IndustrialWeight40 g/m²