VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Catalog # 85025.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- 포장 유형:Plastic bottle
- Pk:100 mL
- 설명:Multi-element calibration standard IV in dil. nitric acid for ICP
Specifications
About this item
23 Elements : 1000 mg/L Ag; Al; B; Ba; Bi; Ca; Cd; Co; Cr; Cu; Fe; Ga; In; K; Li; Mg; Mn; Na; Ni; Pb; Sr; Tl; Zn (HNO₃ 2%)