VWR® ARISTAR®, ICP Multi-Element Solution, 4 Elements in Nitric Acid 7%
제품 번호 85035.180
공급사: VWR Chemicals
CAS 번호:
사양
- 포장 유형:Plastic bottle
- Pk:100 mL
- 설명:ICP multi element solution
사양
이 제품에 관하여
ICP multi-element solution according to USP 232 dietary supplements.
Cd 5 mg/L, Pb 10 mg/L, As 15 mg/L, Hg 15 mg/L in HNO₃ 7%.
Certifications: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.