Orders LinkContactUsLinkComponent
VWR® Ice Buckets, Rectangular, with Lid
VWR® Ice Buckets, Rectangular, with Lid
Supplier:  VWR Collection
Certificates
VWR® Ice Buckets, Rectangular, with Lid
VWR® Ice Buckets, Rectangular, with Lid
8 Options Available Below
Product Details & Documents
Polyurethane foam.
  • Resistant to temperatures from –76 to +70 °C
  • Extremely robust, leakproof, stackable, lightweight
  • Available in two sizes and four colours
  • Stacking option for saving space on working bench
  • Pouring lip design, for easy draining of liquid
  • With lid to reduce the evaporation level of dry ice, water ice with salt slurry
Ideal for storing samples, these buckets maintain hot or cold temperatures of bottles, test tubes, solutions, and cultures. Suitable for use with dry ice, water ice, salt solution. For freezer storage only.
Documents
Product Family Options
Product Information
  • 용량
    Colour
    너비
    깊이
    높이
    Availability
    Price
  • 2,5 L
    Black
    285 mm
    338 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 2,5 L
    Blue
    285 mm
    338 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 2,5 L
    Green
    285 mm
    338 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 2,5 L
    Red
    285 mm
    338 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 4,5 L
    Black
    285 mm
    338 mm
    187 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 4,5 L
    Blue
    285 mm
    338 mm
    187 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 4,5 L
    Green
    285 mm
    338 mm
    187 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 4,5 L
    Red
    285 mm
    338 mm
    187 mm