VWR® ARISTAR® Multi-Component Anion Mix 10, Avantor
Catalog # 458174A
Supplier: VWR Chemicals
CAS Number:
Specifications
- 포장 유형:Plastic bottle
- Pk:1 SET
- 설명:Multi-Component Anion Mix 10
Specifications
About this item
Two solutions - a nitrite standard solution and also a 6 component multi-element standard solution.
Br 50 ppm NO3 50 ppm
Cl 50 ppm PO4 50 ppm
F 25 ppm SO4 50 ppm
NO2 50 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.