Order Entry
Korea, Republic of
ContactUsLinkComponent
headlineimage.jpg
GC Fittings and Accessories
Ensure reproducibility of gas chromatography (GC) results with a large selection of GC fittings and accessories from Avantor®, including chromatography inlet liners, septa, ferrules and more – all specifically designed and optimized for your instrument.
Multiple liner types to prevent adsorption and ensure accurate sample transfer for different GC techniques.
Septa are available in a variety of materials and thicknesses. Ideal for multiple injections due to high resealability.
A wide offering of fittings and ferrules varying in size and material. Also unions, column connectors and column nuts.
Choose from the extensive selection including headspace vials. As well as amber vials for light-sensitive assays.