Order Entry
Korea, Republic of
ContactUsLinkComponent
PFAS KIT 2IN1 548-0011A+548-7006A 1.5ml Snap Ring Vial, ND11, 32 x 11.6mm, clear glass, 1st hydrolytic class, wide opening + 11mm PFAS PE snap ring cap, light-green, with hole, silicon white/transparent polypropylene, SLIT, for PFAS analysis 1 * 1 items
Catalog # VWRI548-7016A
Supplier:  VWR Collection
New Product
PFAS KIT 2IN1 548-0011A+548-7006A 1.5ml Snap Ring Vial, ND11, 32 x 11.6mm, clear glass, 1st hydrolytic class, wide opening + 11mm PFAS PE snap ring cap, light-green, with hole, silicon white/transparent polypropylene, SLIT, for PFAS analysis 1 * 1 items
Catalog # VWRI548-7016A
Supplier:  VWR Collection
New Product