Hose Fittings, Band Clamps, for 6" OD Hose, 25/Pk
MFLX33350-18
Quick and easy assembly without tools.
- Constructed of durable stainless steel
Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.
- Tubing ext.Ø:152,4 mm (6")
- Description:Hose Fittings, Band Clamps, for 6" OD Hose, 25/Pk
- Environmentally Preferable:
- Warranty:1 year
- Pk:25
Frequently Bought Together