Order Entry
Korea, Republic of
ContactUsLinkComponent
Evsco UMB-2 Mounting Brackets for 3/8" and 1/2" NPT(F) elliptic valves, 2/set
Evsco UMB-2 Mounting Brackets for 3/8" and 1/2" NPT(F) elliptic valves, 2/set
Catalog # MFLX98150-51
undefined
Evsco UMB-2 Mounting Brackets for 3/8" and 1/2" NPT(F) elliptic valves, 2/set
Catalog # MFLX98150-51
About this item
Safely mount your plastic elliptical valves.
  • Optimize performance of your Evsco Valves
Constucted of durable polypropylene, these mounting brackets are ideal for making your valve more versitile. These mounting brackets allow for wall, panel, or undermount.
  • Description:
    Mounting Brackets for 3/8" and 1/2" NPT(F) elliptic valves, 2/set
  • Environmentally Preferable:
  • Warranty:
    3 Months
  • Pk:
    1
Frequently Bought Together