Evsco UMB-1 Mounting Brackets for 1/8" and 1/4" NPT(F) elliptic valves, 2/set
Catalog # MFLX98150-50
Supplier: Avantor Fluid Handling
About this item
Safely mount your plastic elliptical valves.
- Optimize performance of your Evsco Valves
Constucted of durable polypropylene, these mounting brackets are ideal for making your valve more versitile. These mounting brackets allow for wall, panel, or undermount.
- Description:Mounting Brackets for 1/8" and 1/4" NPT(F) elliptic valves, 2/set
- Environmentally Preferable:
- Warranty:3 Months
- Pk:1
Frequently Bought Together