Order Entry
Korea, Republic of
ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Straight, Compression to Thread, 1/4" OD x 1/8" NPT(M); 10/PK
Masterflex® Fitting, Polypropylene, Straight, Compression to Thread, 1/4" OD x 1/8" NPT(M); 10/PK
Catalog # MFLX06385-10
undefined
Masterflex® Fitting, Polypropylene, Straight, Compression to Thread, 1/4" OD x 1/8" NPT(M); 10/PK
Catalog # MFLX06385-10
About this item
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/4" OD x 1/8" NPT(M)
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 2):
    ⅛"
  • Description:
    Thread to Compression Adapters, ⅛" NPT(M) to ¼" OD, PP, 10/Pk
  • Seq:
    10
  • Environmentally Preferable:
  • Model:
    06385-10
  • Orifice dimensions:
    5,555 mm (0,21875")
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Fitting material:
    Polypropylene
Frequently Bought Together