Order Entry
Korea, Republic of
ContactUsLinkComponent
Abaque 104884 Lubricant - 1 Gallon
Abaque 104884 Lubricant - 1 Gallon
제품 번호 MFLX75810-89
CE Compliant
undefined
Abaque 104884 Lubricant - 1 Gallon
제품 번호 MFLX75810-89
CE Compliant
About this item
Replacement lubricant for the liquid end coming in four different volumnes.
  • Original lubricant designed to help extend tubing life and to dissipate heat build up.
Optimize performance of your Abaque™ positive displacement pumps.
  • Description:
    Abaque™ lubricant for positive displacement pumps, 3785 ml
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together