Order Entry
Korea, Republic of
ContactUsLinkComponent
Masterflex® Fitting, Polycarbonate, Four-Way, Large-Bore Stopcocks with Male Luer Lock, Non-Sterile; 10/PK
Masterflex® Fitting, Polycarbonate, Four-Way, Large-Bore Stopcocks with Male Luer Lock, Non-Sterile; 10/PK
Catalog # MFLX30600-12
undefined
Masterflex® Fitting, Polycarbonate, Four-Way, Large-Bore Stopcocks with Male Luer Lock, Non-Sterile; 10/PK
Catalog # MFLX30600-12
About this item
Enhance fluid flow with a larger bore than standard luer stopcocks.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Four-way flow pattern
Large-bore adapters are perfect for handling higher flow volumes in process while using connections similar to luer systems.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
  • Flow pattern:
    Four-Way
  • Description:
    Luer fittings, Large-bore stopcocks, male lock, 4-way; 10/pack, Non-sterile
  • Environmentally Preferable:
  • Wetted materials:
    Polycarbonate
  • Warranty:
    Out of box
  • Pk:
    10
  • Inlet port:
    Female Luer
  • Outlet port:
    Male Luer Lock
Frequently Bought Together