Order Entry
Korea, Republic of
ContactUsLinkComponent
Fully Autoclavable 12 Channel Micropipette 30 - 300 µl 1 * 1 items
Catalog # VWRA613-6624
Supplier:  VWR Collection
Fully Autoclavable 12 Channel Micropipette 30 - 300 µl 1 * 1 items
Catalog # VWRA613-6624
Supplier:  VWR Collection