Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
2-Propanol ≥98%, TECHNICAL

2-Propanol ≥98%, TECHNICAL

Canister, 5 L
Catalog # 20922.368
Flammable
Irritant
Supplier:   VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Catalog # 20922.368
Supplier:  VWR Chemicals
CAS Number:  67-63-0
2-Propanol ≥98%, TECHNICAL
2-Propanol ≥98%, TECHNICAL

Canister, 5 L

CAS Number:  67-63-0
Price
₩172,671
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specifications

  • Pack type:
    Canister
  • Pk:
    5 L

Specifications