Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
 

VWR®, Micro cover glasses, rectangular

22×30 mm
Catalog # VWRU48393-026
Supplier:   VWR Collection
VWR®, Micro cover glasses, rectangular
Catalog # VWRU48393-026
Supplier:  VWR Collection
VWR®, Micro cover glasses, rectangular
VWR®, Micro cover glasses, rectangular

22×30 mm

Price
₩23,818
Log in for availability
1 * 28,35 g
Product Details & Documents

These micro cover glasses are made of glass, and each set of numbered slides is uniform in size and thickness.

The No. 1 slides are 0,13 to 0,17 mm in thickness. The No. 1.5 slides are 0,16 to 0,19 mm in thickness.

Specifications
  • L×W:
    22×30 mm
  • Seq:
    01